Order Entry
Switzerland
ContactUsLinkComponent
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Catalog # 34978-2.5L
CAS Number:  
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Catalog # 34978-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay trifluoroacetic acid:
    0.093 - 0.107 %
  • pH (20 °C):
    2.0 - 2.2
  • Calcium (Ca):
    Max. 0.5 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Sodium (Na):
    Max. 2 ppm
  • Transmittance at 210 nm:
    Min. 25 %
  • Transmittance at 230 nm:
    Min. 85 %
  • Transmittance from 254 nm:
    Min. 99 %
  • HPLC Gradient at 210 nm:
    Max. 50 mAU
  • HPLC Gradient at 254 nm:
    Max. 10 mAU
  • LC-MS application:
    complying

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.9982 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:76-05-1
MDL Number:MFCD00011332