Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7732-18-5
MDL Number:MFCD00011332
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.9982 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-580-7
Specification Test Results
- Assay of acetic acid:0.093 - 0.107 %
- pH (20 °C):3.2 - 3.4
- Calcium (Ca):max. 0.5 ppm
- Potassium (K):max. 0.5 ppm
- Magnesium (Mg):max. 0.5 ppm
- Sodium (Na):max. 2 ppm
- Transmittance at 210 nm:min. 20 %
- Transmittance at 230 nm:min. 75 %
- Transmittance from 254 nm:min. 99 %
- HPLC Gradient at 210 nm:max. 50 mAU
- HPLC Gradient at 254 nm:max. 10 mAU
- LC-MS application:complying
Specification Test Results
Frequently Bought Together