
Maleic acid ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:110-16-7
:
Formula:C₄H₄O₄
MDL Number:MFCD00063177
:116.07 g/mol
Merck Index:13,05726
Boiling Pt:256 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1.59 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-742-5
UN:3261
ADR:8,III
:GHS07
Specification Test Results
- Assay (HPLC, area%):Min. 99.0 %
- Assay (acidimetric):Min. 99.0 %
- Identification:Passes test
- Melting point:130 --> 135 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85968.180 spec_VWRC85968.290 spec_VWRC85968.360 spec_VWRC85968.460
Product specification
Frequently Bought Together