ContactUsLinkComponent
Superspher® RP select B, HPLC Columns
 :  Avantor
undefined
Superspher® RP select B, HPLC Columns
 :  Avantor
  • Catalog No:
  • SUPERRPB4250AF
  • SUPERRPB4125AF
  • HICHSUPERRPB4150AF
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 11,5%
  • 11,5%
  • 11,5%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 360 m²/g
  • 360 m²/g
  • 360 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Frequently Bought Together