ContactUsLinkComponent
Superspher® RP18, HPLC Columns
Superspher® RP18, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Superspher® RP18, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • SUPERRP18-4-125AF
  • SUPERRP18425AF
  • HICHSUPERRP184125M
  • SUPERRP184250A
  • SUPERRP18E475A
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)