ContactUsLinkComponent
Sleeve protectors, PP
Sleeve protectors, PP
Catalog # HOIT109760
undefined
Sleeve protectors, PP
Catalog # HOIT109760
Specifications
  • Garment application:
    Laboratory,
    Food
  • Colour:
    White
  • Length:
    475 mm
  • Pk:
    400
Specifications
About this item
These disposable sleeve protectors made of PP provide waterproof protection for light-duty tasks.
  • Lightweight and soft
  • Waterproof and oil-proof
  • Breathable fabric
  • Latex-free
Ideal for use in non-hazardous applications, such as cleaning, maintenance, food processing and handling, manufacturing, hotels and catering.
Weight - 40 g/m²