

Divinylbenzene (mixture of isomers) ≥80% stabilised, Sigma-Aldrich®
Catalog # 8.43986.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:0.005 g/l (25 °C)
- Melting Point:-67 °C
- Molar Mass:130.18 g/mol
- Boiling Point:195 °C (1013 hPa)
- Vapor Pressure:0.9 hPa (20 °C)
- Flash Point:77 °C
- Explosion Limit:1.1 - 6.2 %(V)
- Density:0.91 g/cm3 (20 °C)
- Ignition Point:470 °C
- Synonyms:DVB
- Hill Formula:C10H10
Specification Test Results
About this item
:
:130.19 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
:Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:215-325-5
UN:3082
:GHS07
:GHS09
Storage Temperature:Refrigerator
CAS Number:1321-74-0
Formula:C₁₀H₁₀
MDL Number:MFCD01778656