ContactUsLinkComponent
LiChrospher® RP-8, HPLC Columns
LiChrospher® RP-8, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
LiChrospher® RP-8, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • LISPRP8-5125AF
  • LISPRP8510C5SK
  • LISPRP8-5-10C5
  • LISPRP8-5-125A
  • LISPRP8-5-250A
  • LISPRP8-5250AF
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 12.5%
  • 12,5%
  • 12.5%
  • 12.5%
  • 12.5%
  • 12.5%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)