ContactUsLinkComponent
LiChrospher® RP-18, HPLC Columns
 :  Avantor
undefined
LiChrospher® RP-18, HPLC Columns
 :  Avantor
  • Catalog No:
  • LISPRP18-5250A
  • LISPRP18-5-150AF
  • LISPRP18-5-50AF
  • LISPRP18510C5S
  • LISPRP185300A
  • LISPRP18575AF
  • LISPRP185300AF
  • LISPRP18-5-10C5
  • LISPRP18-5-250AM
  • LISPRP18-5-250AF
  • LISPRP18-5125A
  • LISPRP18-5-125AF
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • 21,0%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Frequently Bought Together