ContactUsLinkComponent
Exsil ODS, HPLC Columns
 :  Avantor
undefined
Exsil ODS, HPLC Columns
 :  Avantor
  • Catalog No:
  • EXODS-5-250AF
  • EXODS-5-10CP3
  • EXODS-5-100AM
  • EXODS1-5-250A
  • EXODS-10-250P
  • EXODS-5-250P
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 11
  • 11
  • 11
  • 7
  • 11
  • 11
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • No
  • Yes
  • Yes
  • Surface area:
  • 200 m²/g
  • 200 m2 /g
  • 200 m²/g
  • 200 m²/g
  • 200 m²/g
  • 200 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Frequently Bought Together