Avantor® Partisil C8, HPLC Columns
: Avantor
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
: * Use with freestanding holder HI-161 and column coupler HI-081, please see “HPLC Guard Column Holders and Accessories”.
- Catalog No:
- P5C8-10C5
- P5C8-10C5-SK
- P5C8-150AS
- P10C8-250P
- P10C8-250A
- P5C8-100P
- P10C8-150AS
- HICHP5C8-75A
- P5C8-100AF
- P5C8-100A
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Frequently Bought Together