Order Entry
Switzerland
ContactUsLinkComponent
Ammonia solution 25%, AR®, Macron Fine Chemicals™
Ammonia solution 25%, AR®, Macron Fine Chemicals™
Certificates
Ammonia solution 25%, AR®, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Assay:
    24 - 28%
  • Residue after Evaporation:
    ≤0.003%
  • Substances Reducing KMnO₄ (as O):
    ≤0.0008%
  • Chloride (Cl):
    ≤0.0001%
  • Phosphate (PO₄):
    ≤0.0002%
  • Total Sulphur (as SO₄):
    ≤0.0003%
  • Carbonate (CO₃):
    ≤0.002%
  • Iron (Fe):
    ≤0.0001%
  • Heavy Metals (as Pb):
    ≤0.001%

Specification Test Results

Related Information

Frequently Bought Together