Polar column test mix, certified reference material, Supelco®
Catalog # 47302.
Supplier: Merck
CAS Number:
Specifications
- Concentration:500 mug/mL each component in dichloromethane
- Pk:1
- Standards:Certified reference material
- Description:Polar Column Test Mix
- Environmentally Preferable:
- Storage temperature:2 - 8 °C
- Pk Info:ampule of 2 mL
Specifications
About this item
Analytes:
2,6-Dimethylaniline; 2,6-Dimethylphenol; Eicosane; Heptadecane; Hexadecane; Octadecane; 1-Octanol; 2-Octanone;
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.