Order Entry
Switzerland
ContactUsLinkComponent
Nucleosides test mix, analytical standard, Supelco®
Nucleosides test mix, analytical standard, Supelco®
Catalog # 47310-U
Supplier:  Merck
CAS Number:  
Nucleosides test mix, analytical standard, Supelco®
Catalog # 47310-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in 1% sodium formate (varied conc.)
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    Nucleosides Test Mix
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    2 - 8 °C
  • Pk Info:
    ampule of 1 mL

Specifications

About this item

Analytes:

Cytidine 50 μg/ml; Guanosine 25 μg/ml; Inosine 25 μg/ml; 1-Methyladenosine 25 μg/ml; 5-Methylcytidine 100 μg/ml; 2′-O-Methylcytidine 20 μg/ml; 3-Methylcytidine methosulfate 100 μg/ml; 7-Methylguanosine 25 μg/ml; 5-Methyluridine 50 μg/ml; β-Pseudouridine 25 μg/ml; 2-Thiocytidine dihydrate 10 μg/ml; Uridine 25 μg/ml