Avantor® Alltima HP C18, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® Alltima HP phase C18 columns are classic reverse phase retention and selectivity.
- Better stability
- High purity
- Excellent column performance
- pH: 1 to 10
Multiple selectivity options for optimising retention, resolution and analysis time. High purity silica helps to eliminate peak tailing problems.
Low column bleed makes the column to be ideal for microbore and other critical analysis.
Specifications
- Catalog No:
- HICH87665
- HICH87681
- HICH87682
- HICH87602
- HICH87603
- HICH87643
- 87683/N
- 87684/N
- HICH87679
- HICH87680
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)