Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Boldine
Catalog # B3916-5G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Boldine
Catalog # B3916-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Standards:
    Analytical standard
  • CAS No.:
    476-70-0
  • Description:
    Boldine
  • Storage conditions:
    RT
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00135040
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,9-Dihydroxy-1,10-dimethoxyaporphine
  • Melting point:
    157 - 164 °C
  • Molecular weight:
    327,37

Specifications

About this item

  • GHS signal: GHS07
Caution: P301 + P312 + P330 - P302 + P352 - P305 + P351 + P338