Trimethyl phosphate solution, NMR reference standard, Supelco®
Catalog # 764353-7IN
Supplier: Merck
CAS Number:
Specifications
- Concentration:25% in chloroform-d (99.8 atom % D), TMS 1 %
- Pk:1
- Standards:NMR reference standard
- Description:Trimethyl phosphate solution
- Description:Trimethyl phosphate solution
- Pk Info:NMR tube size 5 mm x 7 in.