Sucrose solution, NMR reference standard, Supelco®
Catalog # 685887-1EA
Supplier: Merck
CAS Number:
Specifications
- Concentration:2 mM in H2O:D2O (9:1) (99.9 atom % D)
- Pk:1
- Standards:NMR reference standard
- Description:Sucrose solution
- M-Clarity quality level:MQ100
- Description:Sucrose solution
- Pk Info:NMR tube size 5 mm x 8 in.