1,4-Dioxane solution, NMR reference standard, Supelco®
Supplier: Merck
Certificates
About this item
Specifications
- Description:1,4-Dioxane solution
Specifications
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards
Supplier: HAMILTON BONADUZ
Organic reference standard, Astaxanthin, neat, Dr Ehrenstorfer®
Supplier: EHRENSTORFER
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Supelco® PESTANAL® Organic Reference Standard, Isoproturon
Supplier: Merck
Organic reference standard, Trolamine, Ph.Eur., RS
Supplier: EDQM
Certified colour reference standards
Supplier: Lovibond Tintometer
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Melting Point Caffeine 234 - 236,5 C 1 * 1 g
Supplier: A.KRÜSS Optronic