Urea-13C,15N2 solution, (99 atom % 13C, 98 atom % 15N), NMR reference standard, Supelco®
Catalog # 611387-10X7IN
Supplier: Merck
CAS Number:
Specifications
- Concentration:0.1 M in DMSO-d6 (99.9 atom % D)
- Pk:10
- Standards:NMR reference standard
- Description:Urea-13C,15N2 solution, (99 atom % 13C, 98 atom % 15N)
- Description:Urea-13C,15N2 solution
- Pk Info:NMR tube size 5 mm x 7 in.