SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Tiamulin, analytical standard, VETRANAL™, Supelco®
Tiamulin, analytical standard, VETRANAL™, Supelco®
34044-100MG-R
:
Merck
:
Tiamulin, analytical standard, VETRANAL™, Supelco®
34044-100MG-R
:
Merck
:
Pk:
100 mg
Standards:
analytical standard
CAS No.:
55297-95-5
Description:
Tiamulin
M-Clarity quality level:
MQ100
Description:
Tiamulin