Microcystin-LF solution, analytical standard, Supelco®
Catalog # 33762-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:6-9 mug/mL in methanol
- Pk:1 mL
- Standards:analytical standard
- CAS No.:154037-70-4
- Description:Microcystin-LF solution
- M-Clarity quality level:MQ100
- Description:Microcystin-LF solution