Order Entry
Switzerland
ContactUsLinkComponent
 
 
Sulphuric acid ≥95%, AR®, Macron Fine Chemicals™
  3133-46
 :  
 
Sulphuric acid ≥95%, AR®, Macron Fine Chemicals™
  3133-46
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Appearance:
    Clear, Colourless, Oily Liquid
  • Assay:
    ≥95%
  • Residue after Ignition:
    ≤0.001%
  • Substances Reducing KMnO₄ (as SO₂):
    ≤0.001%
  • Nitrate (NO₃):
    ≤0.0002%
  • Chloride (Cl):
    ≤0.0001%
  • Ammonium (NH₄):
    ≤0.0003%
  • Heavy Metals (as Pb):
    ≤0.0002%
  • For Laboratory, Research or Manufacturing Use:
    Passes Test

 

 

 :
 :98.08 g/mol
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589