Micro particles based on melamine resin, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Specifications
- M-Clarity quality level:MQ100