Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Ammonium Standards
Ammonium ion solution for ISE, Supelco®
Ammonium ion solution for ISE, Supelco®
Ammonium ion solution for ISE, Supelco®
09685-250ML
:
Merck
:
Ammonium ion solution for ISE, Supelco®
09685-250ML
:
Merck
:
Concentration:
1000 mg/kg N
Pk:
250 mL
Description:
Ammonium chloride solution