ContactUsLinkComponent
Malate standard for IC, TraceCERT®, Supelco®
  06994-100ML
 :  Merck
 :  
Malate standard for IC, TraceCERT®, Supelco®
  06994-100ML
 :  Merck
 :  
  • Concentration:
    1000 mg/l
  • Pk:
    100 mL
  • Description:
    Malate in water
 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.