Butyrate standard for IC, TraceCERT®, Supelco®
Catalog # 08089-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/l
- Pk:100 mL
- Description:Butyrate in water
- Environmentally Preferable:
Specifications
About this item
Prepared with sodium butyrate and water; filtered through a 0,2 μm membrane filter and stabilised with sodium azide.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.