Titanium Standard for AAS, TraceCERT®, Supelco®
Catalog # 04689-100ML
Supplier: Merck
CAS Number:
Specifications
- Konzentration:1000 mg/L
- Matrix:Salpetersäure verd.
- VE:100 mL
- M-Clarity Qualitätsstufe:MQ300
- Element:Titan
- Symbol:Ti
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034