Ruthenium standard for AAS, analytical standard, Supelco®
Katalognr. 84033-100ML
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Konzentration:1000 mg/L
- Matrix:1 M HCl
- VE:100 mL
- Environmentally Preferable:
- M-Clarity Qualitätsstufe:MQ100
- Element:Ruthenium
- Symbol:Ru
Technische Daten
Über diesen Artikel
Ruthenium standard solution may be used in the determination of ruthenium ions in uranium alloys using atomic absorption spectrophotometry.