Gold Standard for ICP-MS, TraceCERT®, Supelco®
Catalog # 67363-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1 mg/L
- Matrix:5% HCl /tr. HNO₃
- Pk:100 mL
- Environmentally Preferable:
- Element:Gold
- Symbol:Au
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034