Order Entry
Switzerland
ContactUsLinkComponent
Calcium standard for AAS, analytical standard, traceable to BAM, Supelco®
Calcium standard for AAS, analytical standard, traceable to BAM, Supelco®
Catalog # 21049-100ML
Supplier:  Merck
CAS Number:  
Calcium standard for AAS, analytical standard, traceable to BAM, Supelco®
Catalog # 21049-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HCl
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item