
Elastica van Gieson staining kit, Sigma-Aldrich®
Catalog # 1.15974.0002
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:4
- Seq:2740
- Pk Info:4×500 ml
Specifications
About this item
:
Storage Temperature:Refrigerator
UN:2924
ADR:3 (8),
:GHS02
:1