



Methenamine silver plating kit acc. Gomori for histology
Catalog # 1.00820.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre case
- Pk:1
- Seq:0001
- Pk Info:Preparation Capacity 50 slides
Specifications
About this item
:
UN:3316
ADR:9,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
:1