Copper di-ammonium EDTA 0.025 M volumetric solution, for complexometry, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67989-88-2
:
Formula:C₁₀H₂₀CuN₄O₈
MDL Number:MFCD01863036
:387.83 g/mol
:M-Clarity™ quality level = MQ100
Specification Test Results
- Appearance (Colour):Blue
- Appearance (form):Liquid
- Titration(KT) (Molarity):0.024 - 0.026 mol/l
- Density D20/4:1.003 - 1.005
Specification Test Results
Frequently Bought Together