Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzoyl chloride
Benzoyl chloride ≥98.0%, GPR RECTAPUR®
Benzoyl chloride ≥98.0%, GPR RECTAPUR®
:
VWR Chemicals
Benzoyl chloride ≥98.0%, GPR RECTAPUR®
:
VWR Chemicals
:
98-88-4
:
:
C₇H₅ClO
:
MFCD00000653
:
140.57 g/mol
:
1736
:
8,II
:
GHS05
:
GHS07
:
Ambient
Appearance:
Clear liquid
Assay:
≥98.0%
Colour (APHA):
≤50
Identity:
Passes test
SDS_EN_UK_VWRC87035.pdf
sds_VWRC87035
Spec_VWRC87035.290 Benzoyl chloride
Spec_VWRC87035.460 Benzoyl chloride
VWR Chemical Substances for Organic Synthesis
VWR Chemicals Big Packs and Bulks
VWR Chemicals for NMR and UV VIS Spectroscopy
VWR® Anhydrous Solvents
VWR® Green Solvents for Sustainable Chemistry.pdf
Frequently Bought Together
Potassium iodide 99.0-100.5% Ph. Eur.
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
: Merck
L(+)-Tartaric acid ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Magnetic stirring bar retrievers, PTFE coated
: KARTELL
Lewatit® MonoPlus TP 214 ion exchange resin, macroporous 400-1250 μm, Supelco®
: Merck