Order Entry
Switzerland
ContactUsLinkComponent
Diisopropylamine ≥99.0%, GPR RECTAPUR®
Flammable
Corrosive
Irritant
Diisopropylamine ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Corrosive
Irritant
Diisopropylamine ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear yellowish liquid
  • Assay (GC):
    ≥99.0%
  • Relative density (20/4°C):
    0.715 - 0.716
  • Water (K. F.):
    ≤ 0.3 %
  • Identity (IR):
    Passes test

Specification Test Results

Frequently Bought Together