VWR®, TLC Glass Plates, Silica HP, Unmodified Layers
These TLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.
- Specific dyeability and abrasion resistance based on an optimised binder system
- Good wettability for exact colorisation results, even with 100% aqueous detection reagents
Specific surface (BET): ~500 m²/g
Specific pore volume: 0,75 ml/g
Particle size: 5 - 17 µm
Frequently Bought Together