
Nuclear fast red-aluminum sulphate solution 0.1% used in nuclear staining, for histology, Sigma-Aldrich®
Catalog # 1.00121.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 ml
- Seq:2725
- Pk Info:Preparation Capacity 2000 - 2500 slides
Specifications
Specification Test Results
- Suitability for microscopy:passes test
- Nuclei:dark red
- Cytoplasm:light red
- Acid mucosubstances:light blue
Specification Test Results
About this item
:
Density:1.04…1.06 g/cm3 (20 °C)
:GHS05
Storage Temperature:Ambient
:1