Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Standards
Organic Standards
Organic reference standard, Zearalenone, Supelco®
Organic reference standard, Zearalenone, Supelco®
32939-5MG
:
Merck
:
Organic reference standard, Zearalenone, Supelco®
32939-5MG
:
Merck
:
Pk:
5 mg
CAS No.:
17924-92-4
Description:
Zearalenone
M-Clarity quality level:
MQ200
Formula:
C₁₈H₂₂O₅
Storage temperature:
−20 °C
Description:
Zearalenone
Form:
Neat