Organic reference standard, Trichothecene Mix solution, analytical standard, Supelco®
Catalog # 32926-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- Description:Trichothecene Mix solution
- M-Clarity quality level:MQ100
- Storage temperature:−20 °C
- Description:Trichothecene Mix solution