Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Trans,Trans-Farnesyl Pyrophosphate Ammonium Salt
Supelco® Organic Reference Standard, Trans,Trans-Farnesyl Pyrophosphate Ammonium Salt
Catalog # 44722-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Trans,Trans-Farnesyl Pyrophosphate Ammonium Salt
Catalog # 44722-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • Description:
    trans,trans-Farnesyl pyrophosphate ammonium salt
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt; Farnesyl diphosphate ammonium salt
  • Formula:
    C₁₅H₂₈O₇P₂ ·xNH₃
  • Molecular weight:
    382,33 (free acid basis)
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.