Organic reference standard, Sodium metabisulfite, analytical standard, Supelco®
Catalog # 08982-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:Analytical standard
- CAS No.:7681-57-4
- Description:Sodium metabisulfite, ≥98,0% (iodometric, RT)
- Melting point:>300 °C (lit.)
- Formula:Na₂S₂O₅
- Description:Sodium metabisulfite
Specifications
About this item
Sodium metabisulphite is primarily used as an antioxidant and antimicrobial agent in a wide variety of foods and drugs. It is also used as the main additive in the prevention of melanosis.
Sodium metabisulphite may be used as an antioxidant which when added to anesthetic formulations serves to minimize epinephrine oxidation in the quantification of epinephrine in dental anesthetic solutions by capillary electrophoresis. Sodium metabisulphite may also be used as a standard in the determination of catecholamines in human plasma by high-performance liquid chromatography with electrochemical detection.
Analysis Note - metal trace analysis, corresponds to requirements