- Pk:10 mg
- Standards:Analytical standard
- CAS No.:37271-16-2
- Description:Sennoside C
- Environmentally Preferable:
- Storage conditions:2…8 °C
- Hazard class description:H302
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Molecular weight:848,76
- Flash point:344,4 °C
: P264 - P270 - P301 + P312 - P501