Supelco® Organic Reference Standard, Schisandrin A
90139-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:61281-38-7
- Description:Schisandrin A
- Environmentally Preferable:
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD09026934
- M-Clarity quality level:MQ100
- Synonym(s):Deoxyschisandrin; Deoxyschizandrin; Schizandrin A
- Molecular weight:416,51