Organic reference standard, Pyrrole, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
Pyrrole is used as one of the building blocks of biochemical macrocycles such as chlorins, porphyrins and corrins. It is also used for many technologically relevel applications such as in organometallic complexes and precursor molecules for self-assembled monolayers on metal surfaces.
Specifications
- CAS No.:109-97-7
- Melting point:–23 °C
- Formula:C₄H₅N
- Storage temperature:2...8 °C
- Description:Pyrrole
- Flash point:36 °C
- Boiling point:131 °C
Specifications
Frequently Bought Together