Organic reference standard, Ponceau 6R, analytical standard, Supelco®
Catalog # 96365-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:2766-77-0
- Description:Ponceau 6R, ≥98,0% (HPLC)
- Melting point:>300 °C (lit.)
- Formula:C₂₀H₁₂N₂Na₂O₇S₂
- Description:Ponceau 6R
- Form:Neat
Specifications
About this item
Ponceau 6R is a red azo dye which is widely used as a food colorant and also used in histology staining. It is soluble in water and slightly soluble in ethanol.
Ponceau 6R (P6R) has been used as a reference standard for the determination of synthetic food and beverages colorants by high-performance liquid chromatography coupled with diode-array detector and as a model compound for detailed study of the mechanism of an azo-dye degradation by soybean peroxidase enzyme (SBP). It may be used as a reference standard in the determination of ponceau 6R in drinks and candy samples using reversed-phase high performance liquid chromatography (RP-HPLC).
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.