Organic reference standard, Ponceau 4R, analytical standard, Supelco®
Catalog # 18137-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:2611-82-7
- Description:Ponceau 4R, ≥99,0% (HPLC)
- Formula:C₂₀H₁₁N₂Na₃O₁₀S₃
- Description:Ponceau 4R
- Form:Neat
Specifications
About this item
Ponceau 4R is a synthetic red azo dye and is used as a food colorant. It is used in range of alcoholic and non-alcoholic beverages, food stuffs, preserved fruits and more. It exists as trisodium-2-hydroxy-1-(4-sulfonato-1-naphthylazo)-6,8-naphthalenedisulphonate.
Ponceau 4R was used in the determination of contents of several additives in different non-alcoholic beverages using HPLC. It was used in understanding the genotoxicity of colorants. It was also used in preparing a falsified drink in the analysis of methanol-ethanol mixture using biosensors amperometric system.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.