Organic reference standard, Orange II sodium salt, analytical standard, Supelco®
Catalog # 69143-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:633-96-5
- Description:Orange II sodium salt
- M-Clarity quality level:MQ100
- Formula:C₁₆H₁₁N₂NaO₄S
- Description:Orange II sodium salt
- Form:Neat
Specifications
About this item
Orange II sodium salt can undergo oxidation by hydrogen peroxide, which is catalysed by ferric ion complexed to tetra amido macrocyclic ligands(FeIII–TAML activators) to give out carbon dioxide, carbon monoxide, small aliphatic carboxylic acids and phthalic acid as the major products.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.