Supelco® Organic Reference Standard, Neoandrographolide
Catalog # 49879-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:27215-14-1
- Description:Neoandrographolide
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD07779130
- M-Clarity quality level:MQ100
- Synonym(s):ent-19-Hydroxy-8(17),13-labdadien-16,15-olide 19-O-β-ᴅ-glucopyranoside; ᴅ-Glucopyranosyloxy)methyl]decahydro-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5H)-furanone
- Molecular weight:480,59