Supelco® Organic Reference Standard, Naringin
Catalog # 91842-500MG
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mg
- Standards:Analytical standard
- CAS No.:10236-47-2
- Description:Naringin
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00148888
- M-Clarity quality level:MQ100
- Synonym(s):4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside; Naringenin 7-neohesperidoside; Naringenine-7-rhamnosidoglucoside; Naringoside
- Molecular weight:580,53
Specifications
About this item
- GHS signal: GHS07
Caution: P301 + P312 + P330